1-(2-Isopropylphenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89198-500MG 89198-1G 89198-5G 89198-25G
77335-050EA 161.71 USD
77335-050 77346-042 77371-084 77391-602
1-(2-Isopropylphenyl)cyclohexanol ≥95%
1-(2-Isopropylphenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₅H₂₂O
MW: 218.335 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771854
CAS Number: 1875124-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR