1-(2-Chloro-5-methylphenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89186-500MG 89186-1G 89186-5G 89186-25G
77335-048EA 250.29 USD
77335-048 77359-150 77371-082 77391-600
1-(2-Chloro-5-methylphenyl)cyclohexanol ≥97%
1-(2-Chloro-5-methylphenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClO
MW: 224.726 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771853
CAS Number: 1995682-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR