1-(2-Bromo-6-fluorophenyl)cyclohexanol

Supplier: AOBChem USA

Warning

89144-500MG 89144-1G 89144-5G 89144-25G
77335-040EA 161.71 USD
77335-040 77346-036 77371-074 77391-592
1-(2-Bromo-6-fluorophenyl)cyclohexanol
1-(2-Bromo-6-fluorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄BrFO
MW: 273.141 g/mol
Storage Temperature: Ambient
CAS Number: 952708-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR