1-(3,5-Difluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89143-500MG 89143-1G 89143-5G
77335-038EA 318.27 USD
77335-038 77359-146 77383-106
1-(3,5-Difluorophenyl)cyclohexanol ≥95%
1-(3,5-Difluorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O
MW: 212.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222219
CAS Number: 1340512-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR