1-(2,4-Dichlorophenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

89139-500MG 89139-1G 89139-5G 89139-25G
77335-036EA 161.71 USD
77335-036 77346-034 77371-072 77391-590
1-(2,4-Dichlorophenyl)cyclohexanol ≥97%
1-(2,4-Dichlorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂O
MW: 245.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771841
CAS Number: 40254-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR