1-(3,4-Dichlorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89138-500MG 89138-1G 89138-5G 89138-25G
77335-034EA 120.51 USD
77335-034 77343-850 77371-070 77383-104
1-(3,4-Dichlorophenyl)cyclohexanol ≥95%
1-(3,4-Dichlorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂O
MW: 245.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16786807
CAS Number: 40254-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR