1-(3-Chlorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

89112-500MG 89112-1G 89112-5G 89112-25G
77335-026EA 101.97 USD
77335-026 77343-024 77371-062 77383-098
1-(3-Chlorophenyl)cyclohexanol ≥95%
1-(3-Chlorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClO
MW: 210.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16111853
CAS Number: 19920-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR