1,3-Dibromo-2-methoxy-5-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

88977-500MG 88977-1G 88977-5G 88977-25G
77335-024EA 176.13 USD
77335-024 77346-568 77371-060 77391-582
1,3-Dibromo-2-methoxy-5-(methoxymethyl)benzene ≥97%
1,3-Dibromo-2-methoxy-5-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂O₂
MW: 309.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771746
CAS Number: 2969233-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR