2-Bromo-4-chloro-3-ethoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

88693-500MG 88693-1G 88693-25G 88693-5G
77335-020EA 300.76 USD
77335-020 77359-138 77391-578 77383-096
2-Bromo-4-chloro-3-ethoxy-1-methylbenzene ≥97%
2-Bromo-4-chloro-3-ethoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771642
CAS Number: 2383486-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR