2-Bromo-4-chloro-3-methoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

88692-500MG 88692-1G 88692-5G
77335-018EA 331.66 USD
77335-018 77359-136 77383-094
2-Bromo-4-chloro-3-methoxy-1-methylbenzene ≥97%
2-Bromo-4-chloro-3-methoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD22414753
CAS Number: 1785158-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR