3-Bromo-5-(ethylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

88514-500MG 88514-1G 88514-5G 88514-25G
77335-016EA 182.31 USD
77335-016 77347-058 77371-056 77391-576
3-Bromo-5-(ethylthio)benzoic acid ≥97%
3-Bromo-5-(ethylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂S
MW: 261.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22483748
CAS Number: 1592949-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR