3-(Ethylthio)-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

88506-1G 88506-5G 88506-25G
77342-058EA 111.24 USD
77342-058 77371-052 77383-090
3-(Ethylthio)-5-fluorobenzoic acid ≥97%
3-(Ethylthio)-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22483652
CAS Number: 453566-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR