3-Chloro-5-(ethylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

88498-500MG 88498-1G 88498-5G 88498-25G
77335-010EA 136.99 USD
77335-010 77344-642 77371-046 77391-572
3-Chloro-5-(ethylthio)benzoic acid ≥97%
3-Chloro-5-(ethylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD22483695
CAS Number: 1822666-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR