3-(Ethylthio)-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

88492-500MG 88492-1G 88492-5G 88492-25G
77335-006EA 93.73 USD
77335-006 77342-494 77371-042 77383-084
3-(Ethylthio)-5-(trifluoromethyl)benzaldehyde ≥97%
3-(Ethylthio)-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃OS
MW: 234.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771547
CAS Number: 2901105-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR