1-Iodo-4-isobutoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

88441-500MG 88441-1G 88441-5G 88441-25G
77335-004EA 211.15 USD
77335-004 77359-134 77371-038 77391-570
1-Iodo-4-isobutoxy-2-methylbenzene ≥97%
1-Iodo-4-isobutoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅IO
MW: 290.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32649098
CAS Number: 2748187-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR