(4-Isobutoxy-2-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

88440-500MG 88440-1G 88440-5G 88440-25G
77335-002EA 144.2 USD
77335-002 77345-028 77371-036 77391-568
(4-Isobutoxy-2-methylphenyl)(methyl)sulfane ≥95%
(4-Isobutoxy-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₂H₁₈OS
MW: 210.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771511
CAS Number: 2901066-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR