9-propyl-9H-carbazole-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

88415-500MG 88415-1G 88415-5G 88415-25G
77334-996EA 146.26 USD
77334-996 77345-184 77371-030 77391-562
9-propyl-9H-carbazole-3-carboxylic acid ≥97%
9-propyl-9H-carbazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₂
MW: 253.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14711162
CAS Number: 892218-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR