2-Bromo-4-(9H-carbazol-9-yl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

88305-500MG 88305-1G 88305-5G
77334-994EA 324.45 USD
77334-994 77359-126 77383-076
2-Bromo-4-(9H-carbazol-9-yl)benzoic acid ≥97%
2-Bromo-4-(9H-carbazol-9-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₂BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD34771434
CAS Number: 2888526-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR