4-(9H-carbazol-9-yl)-3-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

88301-500MG 88301-1G 88301-5G
77338-968EA 536.63 USD
77338-968 77359-124 77383-074
4-(9H-carbazol-9-yl)-3-chlorobenzaldehyde ≥97%
4-(9H-carbazol-9-yl)-3-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₉H₁₂ClNO
Storage Temperature: Ambient
MDL Number: MFCD34771433
CAS Number: 2901109-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR