(2,6-Dichloro-3-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

88245-500MG 88245-1G 88245-5G 88245-25G
77334-992EA 82.4 USD
77334-992 77342-170 77371-028 77383-068
(2,6-Dichloro-3-iodophenyl)(methyl)sulfane ≥97%
(2,6-Dichloro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅Cl₂IS
MW: 318.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771419
CAS Number: 2706159-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR