Methyl 2,4-dichloro-3-(methylthio)benzoate ≥95%

Supplier: AOBChem USA

Warning

88243-500MG 88243-1G 88243-5G 88243-25G
77334-988EA 120.51 USD
77334-988 77343-848 77371-024 77383-064
Methyl 2,4-dichloro-3-(methylthio)benzoate ≥95%
Methyl 2,4-dichloro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂S
MW: 251.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771417
CAS Number: 2918860-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR