1,3-Dibromo-2-ethoxy-5-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

88001-500MG 88001-1G 88001-5G 88001-25G
77334-986EA 121.54 USD
77334-986 77343-886 77371-022 77383-062
1,3-Dibromo-2-ethoxy-5-nitrobenzene ≥97%
1,3-Dibromo-2-ethoxy-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD34771337
CAS Number: 31106-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR