4-Bromo-1-isopropoxy-2-nitrobenzene ≥95%

Supplier: AOBChem USA

Warning

87967-1G 87967-5G 87967-25G
77340-842EA 60.77 USD
77340-842 77371-018 77383-058
4-Bromo-1-isopropoxy-2-nitrobenzene ≥95%
4-Bromo-1-isopropoxy-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD14652100
CAS Number: 383869-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR