1-(3,6-Dibromo-2-fluorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

87957-500MG 87957-1G 87957-5G 87957-25G
77334-984EA 254.41 USD
77334-984 77359-118 77371-016 77391-560
1-(3,6-Dibromo-2-fluorophenyl)-2-fluoroethanone ≥95%
1-(3,6-Dibromo-2-fluorophenyl)-2-fluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄Br₂F₂O
MW: 313.922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771316
CAS Number: 2918847-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR