Methyl 2-methyl-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
87834-500MG
87834-1G
87834-5G
87834-25G
77334-980EA
186.43
USD
77334-980
77359-114
77371-012
77391-556
Methyl 2-methyl-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Methyl 2-methyl-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₅H₂₀BNO₆ MW: 321.133 g/mol Storage Temperature: Ambient |
CAS Number:
1547162-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...