3-Bromo-2-iodo-5-nitrotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-iodo-3-methyl-5-nitrobenzene

87644-500MG 87644-1G 87644-5G 87644-25G
77334-976EA 107.12 USD
77334-976 77343-140 77371-008 77383-050
3-Bromo-2-iodo-5-nitrotoluene ≥97%
3-Bromo-2-iodo-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrINO₂
Storage Temperature: Ambient
MDL Number: MFCD11846022
CAS Number: 1160573-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR