6-Methyl-2,3-dihydrobenzo[b][1,4]dioxine ≥97%
Supplier: AOBChem USA
|
Warning
|
87630-500MG
87630-1G
87630-5G
87630-25G
77334-974EA
146.26
USD
77334-974
77345-182
77371-006
77391-552
6-Methyl-2,3-dihydrobenzo[b][1,4]dioxine ≥97%
6-Methyl-2,3-dihydrobenzo[b][1,4]dioxine
Precaución: for Research Use Only
Formula:
C₉H₁₀O₂ MW: 150.174 g/mol Storage Temperature: Ambient |
CAS Number:
33632-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...