1-(2,4-Dichloro-3-iodophenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
87615-500MG
87615-1G
87615-5G
87615-25G
77334-972EA
89.61
USD
77334-972
77342-374
77371-004
77383-048
1-(2,4-Dichloro-3-iodophenyl)ethanone ≥95%
1-(2,4-Dichloro-3-iodophenyl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₅Cl₂IO MW: 314.935 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34771265 CAS Number: 2383436-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...