3-Acetyl-2,6-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

87610-1G 87610-5G 87610-25G
77341-448EA 86.52 USD
77341-448 77371-000 77383-046
3-Acetyl-2,6-dichlorobenzoic acid ≥97%
3-Acetyl-2,6-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₃
MW: 233.048 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771260
CAS Number: 2901086-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR