1,3-Dichloro-4-ethyl-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

87604-500MG 87604-1G 87604-5G 87604-25G
77334-968EA 124.63 USD
77334-968 77344-068 77370-996 77383-042
1,3-Dichloro-4-ethyl-2-iodobenzene ≥95%
1,3-Dichloro-4-ethyl-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇Cl₂I
MW: 300.952 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771255
CAS Number: 2711146-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR