(2,6-Dichloro-3-ethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

87603-500MG 87603-1G 87603-5G 87603-25G
77334-966EA 126.69 USD
77334-966 77344-196 77370-994 77383-040
(2,6-Dichloro-3-ethylphenyl)(methyl)sulfane ≥97%
(2,6-Dichloro-3-ethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂S
MW: 221.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771254
CAS Number: 2710812-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR