2,6-Dichloro-3-ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

87600-500MG 87600-1G 87600-5G 87600-25G
77334-964EA 130.81 USD
77334-964 77344-362 77370-992 77391-548
2,6-Dichloro-3-ethylbenzoic acid ≥97%
2,6-Dichloro-3-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MW: 219.065 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34771252
CAS Number: 2709799-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR