1-(2,4-Dichloro-5-nitrophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

87592-1G 87592-5G 87592-25G
77340-840EA 60.77 USD
77340-840 77370-988 77383-038
1-(2,4-Dichloro-5-nitrophenyl)ethanone ≥97%
1-(2,4-Dichloro-5-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅Cl₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD00587974
CAS Number: 61671-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR