2-Bromo-1-fluoro-3,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

87302-500MG 87302-1G 87302-5G 87302-25G
77334-958EA 181.28 USD
77334-958 77346-878 77370-984 77391-542
2-Bromo-1-fluoro-3,4-dimethoxybenzene ≥97%
2-Bromo-1-fluoro-3,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738430
CAS Number: 1637490-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR