3-Bromo-4-fluoro-5-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

87300-500MG 87300-1G 87300-5G
77335-740EA 363.59 USD
77335-740 77359-104 77383-036
3-Bromo-4-fluoro-5-hydroxybenzaldehyde ≥97%
3-Bromo-4-fluoro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
MW: 219.008 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28613067
CAS Number: 1779914-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR