3-Ethoxy-2,4-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

87109-500MG 87109-1G 87109-5G
77334-952EA 332.69 USD
77334-952 77359-096 77383-032
3-Ethoxy-2,4-dimethoxybenzaldehyde ≥97%
3-Ethoxy-2,4-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD34771093
CAS Number: 69505-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR