3-Methyl-4-phenoxyaniline ≥97%

Supplier: AOBChem USA

Warning

87051-500MG 87051-1G 87051-25G 87051-5G
77334-950EA 299.73 USD
77334-950 77359-092 77391-536 77383-028
3-Methyl-4-phenoxyaniline ≥97%
3-Methyl-4-phenoxyaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD08688318
CAS Number: 191284-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR