2,3-Dichloro-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

87005-500MG 87005-1G 87005-5G 87005-25G
77334-944EA 120.51 USD
77334-944 77343-846 77370-974 77383-026
2,3-Dichloro-4-phenylpyridine ≥97%
2,3-Dichloro-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₇Cl₂N
MW: 224.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787394
CAS Number: 1255311-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR