(3-Chloro-5-isopropylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

86754-500MG 86754-1G 86754-5G
77337-996EA 456.29 USD
77337-996 77359-086 77383-022
(3-Chloro-5-isopropylphenyl)boronic acid ≥97%
(3-Chloro-5-isopropylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BClO₂
MW: 198.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31726378
CAS Number: 2919766-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR