1-Bromo-3-chloro-2-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

86664-500MG 86664-1G 86664-5G 86664-25G
77334-938EA 141.11 USD
77334-938 77344-948 77370-968 77391-528
1-Bromo-3-chloro-2-methoxy-5-methylbenzene ≥97%
1-Bromo-3-chloro-2-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD22414754
CAS Number: 1824192-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR