2-(5-Methoxy-4-methylpyridin-3-yl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

86490-500MG 86490-1G 86490-5G 86490-25G
77334-936EA 223.51 USD
77334-936 77359-080 77370-966 77391-526
2-(5-Methoxy-4-methylpyridin-3-yl)propan-2-ol ≥97%
2-(5-Methoxy-4-methylpyridin-3-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD34770711
CAS Number: 2284050-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR