1-(5-Methoxy-4-methylpyridin-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

86489-500MG 86489-1G 86489-5G 86489-25G
77334-934EA 226.6 USD
77334-934 77359-078 77370-964 77391-524
1-(5-Methoxy-4-methylpyridin-3-yl)ethanone ≥97%
1-(5-Methoxy-4-methylpyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD34770710
CAS Number: 1935026-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR