3-Methoxy-4-methyl-5-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

86486-500MG 86486-1G 86486-5G 86486-25G
77334-930EA 156.56 USD
77334-930 77345-634 77370-962 77391-520
3-Methoxy-4-methyl-5-(methylthio)pyridine ≥95%
3-Methoxy-4-methyl-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NOS
MW: 169.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770708
CAS Number: 77903-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR