(3-(Difluoromethoxy)-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

86440-500MG 86440-1G
77340-580EA 2651.22 USD
77340-580 77359-074
(3-(Difluoromethoxy)-5-methylphenyl)(methyl)sulfane ≥97%
(3-(Difluoromethoxy)-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀F₂OS
MW: 204.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770707
CAS Number: 2901066-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR