(3-Isopropoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

86434-500MG 86434-1G 86434-5G
77338-174EA 466.59 USD
77338-174 77359-070 77383-012
(3-Isopropoxy-5-methylphenyl)(methyl)sulfane ≥97%
(3-Isopropoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₆OS
MW: 196.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34770704
CAS Number: 2901103-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR