1-Ethoxy-3-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

86429-500MG 86429-1G 86429-5G
77334-928EA 347.11 USD
77334-928 77359-068 77383-010
1-Ethoxy-3-iodo-5-methylbenzene ≥97%
1-Ethoxy-3-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD34770700
CAS Number: 2918916-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR