(3-Ethoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

86428-500MG 86428-1G 86428-5G
77335-738EA 363.59 USD
77335-738 77359-066 77383-008
(3-Ethoxy-5-methylphenyl)(methyl)sulfane ≥97%
(3-Ethoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD34770699
CAS Number: 2918838-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR