1-(3-Methoxy-5-methylphenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
86426-500MG
86426-1G
86426-5G
86426-25G
77334-926EA
87.55
USD
77334-926
77342-314
77370-960
77383-006
1-(3-Methoxy-5-methylphenyl)ethanone ≥97%
1-(3-Methoxy-5-methylphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₂ MW: 164.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17011766 CAS Number: 43113-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...