1-(3-Bromo-5-(methylthio)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

86419-500MG 86419-1G 86419-5G 86419-25G
77334-924EA 182.31 USD
77334-924 77347-056 77370-958 77391-518
1-(3-Bromo-5-(methylthio)phenyl)ethanone ≥95%
1-(3-Bromo-5-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrOS
MW: 245.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34535367
CAS Number: 1779765-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR