(3-Bromo-5-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

86418-500MG 86418-1G 86418-5G 86418-25G
77334-922EA 174.07 USD
77334-922 77346-472 77370-956 77391-516
(3-Bromo-5-iodophenyl)(methyl)sulfane ≥97%
(3-Bromo-5-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₆BrIS
Storage Temperature: Ambient
MDL Number: MFCD22488912
CAS Number: 1822986-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR